Gene Transcripts

Gene Symbol
LCE2C
Entrez Gene ID
353140
Gene Name
late cornified envelope 2C
Gene Aliases
LEP11
Chromosome Location
Chr. 1: 152675315 - 152676574 on Build GRCh38
UniGene ID
Hs.553713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178429.3 NP_848516.1 1 386
GenBank mRNA BC125197.1 - 1 386

View Full Product Details