Gene Transcripts

Gene Symbol
FAM212A
Entrez Gene ID
389119
Gene Name
family with sequence similarity 212 member A
Gene Aliases
C3orf54, INKA1
Chromosome Location
Chr. 3: 49803130 - 49805030 on Build GRCh38
UniGene ID
Hs.86674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203370.1 NP_976248.1 2 937
XM_017006372.1 XP_016861861.1 2 981
GenBank mRNA BC012170.1 - 2 937

View Full Product Details