Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_015356.2 | 3 | 294 | |||
NR_033664.1 | 2 | 223 | ||||
NR_033665.1 | 2 | 223 | ||||
GenBank mRNA | AY312364.1 | - | 3 | 297 | ||
AY312365.1 | - | LIT3317 AAA1 |
3 | 292 | ||
AY312366.1 | - | LIT3326 AAA1 |
3 | 188 | ||
AY312367.1 | - | LIT3327 AAA1 |
2 | 223 | ||
AY312368.1 | - | LIT3328 AAA1 |
2 | 223 | ||
AY312369.1 | - | LIT3332 AAA1 |
2 | 223 | ||
AY312370.1 | - | LIT3333 AAA1 |
2 | 108 | ||
AY312371.1 | - | LIT3334 AAA1 |
2 | 223 | ||
AY312372.1 | - | LIT3335 AAA1 |
2 | 223 | ||
AY312373.1 | - | LIT3331 AAA1 |
3 | 188 | ||
AY321515.1 | - | 2 | 104 | |||
AY321516.1 | - | 2 | 104 | |||
BC031961.1 | - | 3 | 275 | |||
BC130477.1 | - | 2 | 108 | |||
BX649082.1 | - | 2 | 186 | |||
U85653.1 | - | 2 | 186 |