Gene Transcripts

Gene Symbol
RSPO4
Entrez Gene ID
343637
Gene Name
R-spondin 4
Gene Aliases
C20orf182, CRISTIN4
Chromosome Location
Chr. 20: 958452 - 1002264 on Build GRCh38
UniGene ID
Hs.444980
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029871.3 NP_001025042.2 5 1226
NM_001040007.2 NP_001035096.1 4 1040
GenBank mRNA AK122609.1 - 5 1223

View Full Product Details