Gene Transcripts

Gene Symbol
MESDC2
Entrez Gene ID
23184
Gene Name
mesoderm development candidate 2
Gene Aliases
BOCA, MESD
Chromosome Location
Chr. 15: 80947326 - 80989878 on Build GRCh38
UniGene ID
Hs.578450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015154.2 NP_055969.1 3 1581
GenBank mRNA AK026606.1 - 3 1509
AY359110.1 - 3 1567
D42039.2 - 3 1497

View Full Product Details