Gene Transcripts

Gene Symbol
MESDC2
Entrez Gene ID
23184
Gene Name
mesoderm development candidate 2
Gene Aliases
BOCA, MESD
Chromosome Location
Chr. 15: 80947326 - 80989878 on Build GRCh38
UniGene ID
Hs.578450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015154.2 NP_055969.1 3 2652
GenBank mRNA D42039.2 - 3 2568

View Full Product Details