Gene Transcripts

Gene Symbol
CRTAM
Entrez Gene ID
56253
Gene Name
cytotoxic and regulatory T-cell molecule
Gene Aliases
CD355
Chromosome Location
Chr. 11: 122838419 - 122872643 on Build GRCh38
UniGene ID
Hs.159523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019604.3 NP_062550.2 3 337
GenBank mRNA AF001622.1 - 3 288
AK313578.1 - 3 335
BC070266.1 - 3 337 30418882

View Full Product Details