Gene Transcripts

Gene Symbol
Snapin
Entrez Gene ID
295217
Gene Name
SNAP-associated protein
Gene Aliases
Snapap
Chromosome Location
Chr. 2: 189878159 - 189880743 on Build Rnor_6.0
UniGene ID
Rn.3815
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025648.1 NP_001020819.1 2 1088
NM_001170576.1 NP_001164047.1 4 1005
GenBank mRNA BC098864.1 - 2 1088
FQ218987.1 - 2 695

View Full Product Details