Gene Transcripts

Gene Symbol
Rhox7
Entrez Gene ID
298353
Gene Name
reproductive homeobox 7
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.144870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025654.1 NP_001020825.1 1 57
GenBank mRNA DQ058654.1 - 1 57

View Full Product Details