Gene Transcripts

Gene Symbol
Phf20l1
Entrez Gene ID
314964
Gene Name
PHD finger protein 20-like 1
Gene Aliases
Chromosome Location
Chr. 7: 107406948 - 107445219 on Build Rnor_6.0
UniGene ID
Rn.22288
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271439.1 NP_001258368.1 778
XM_008765533.1 XP_008763755.1 6 573
XM_008765534.1 XP_008763756.1 5 500
XM_008765536.1 XP_008763758.1 6 574
XM_008765538.1 XP_008763760.1 3 201
GenBank mRNA BC096896.1 - 6 574

View Full Product Details