Gene Transcripts

Gene Symbol
Tfeb
Entrez Gene ID
316214
Gene Name
transcription factor EB
Gene Aliases
Tcfeb
Chromosome Location
Chr. 9: 15208141 - 15263966 on Build Rnor_6.0
UniGene ID
Rn.22098
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025707.1 NP_001020878.1 4, 5 911
XM_006244440.2 XP_006244502.1 5, 6 1840
XM_006244442.1 XP_006244504.1 5, 6 830
XM_006244446.2 XP_006244508.1 5, 6 2101
GenBank mRNA BC099102.1 - 4, 5 911

View Full Product Details