Gene Transcripts

Gene Symbol
Proca1
Entrez Gene ID
497959
Gene Name
protein interacting with cyclin A1
Gene Aliases
Chromosome Location
Chr. 10: 65454658 - 65469682 on Build Rnor_6.0
UniGene ID
Rn.230966
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025758.1 NP_001020929.1 4 774
XM_008768093.1 XP_008766315.1 4 680
GenBank mRNA BC098036.1 - 4 774

View Full Product Details