Gene Transcripts

Gene Symbol
Pcdhga8
Entrez Gene ID
364843
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDHGB5
Chromosome Location
Chr. 18: 30874873 - 30971113 on Build Rnor_6.0
UniGene ID
Rn.8628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037156.1 NP_001032233.1 1 1592
GenBank mRNA AY574019.1 - 1 1592

View Full Product Details