Gene Transcripts

Gene Symbol
Nxf7
Entrez Gene ID
501621
Gene Name
nuclear RNA export factor 7
Gene Aliases
Chromosome Location
Chr. X: 106049075 - 106066342 on Build Rnor_6.0
UniGene ID
Rn.24735
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037216.1 NP_001032293.1 22 2219
XM_006257254.2 XP_006257316.1 22 2252
XM_008773431.1 XP_008771653.1 22 2790
GenBank mRNA BC100151.1 - 22 2219
FQ229456.1 - 11 1046

View Full Product Details