Gene Transcripts

Gene Symbol
Mrgpra1
Entrez Gene ID
233221
Gene Name
MAS-related GPR, member A1
Gene Aliases
MrgA1
Chromosome Location
Chr. 7: 47334875 - 47354240 on Build GRCm38
UniGene ID
Mm.389895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153095.2 NP_694735.2 2 719
GenBank mRNA AY042191.1 - 2 719
BC126873.1 - 1 649 40044758

View Full Product Details