Gene Transcripts

Gene Symbol
Olfr58
Entrez Gene ID
18358
Gene Name
olfactory receptor 58
Gene Aliases
IG6, MOR146-3, MOR146-7P
Chromosome Location
Chr. 9: 19780283 - 19784064 on Build GRCm38
UniGene ID
Mm.248950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011001.2 NP_035131.1 2 871
GenBank mRNA AF102536.1 - 1 427

View Full Product Details