Gene Transcripts
- Gene Symbol
- SLC12A9
- Entrez Gene ID
- 56996
- Gene Name
- solute carrier family 12 member 9
- Gene Aliases
- CCC6, CIP1, WO3.3, hCCC6
- Chromosome Location
- Chr. 7: 100852719 - 100867012 on Build GRCh38
- UniGene ID
- Hs.521087
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267812.1 |
NP_001254741.1
|
11, 12 |
1704 |
|
NM_001267814.1 |
NP_001254743.1
|
9, 10 |
1441 |
|
NM_020246.3 |
NP_064631.2
|
11, 12 |
1704 |
|
XM_005250502.2 |
XP_005250559.1
|
9, 10 |
1440 |
|
XM_005250504.4 |
XP_005250561.1
|
5, 6 |
675 |
|
XM_006716054.2 |
XP_006716117.1
|
9, 10 |
1422 |
|
XM_006716055.2 |
XP_006716118.1
|
9, 10 |
1277 |
|
XM_011516414.1 |
XP_011514716.1
|
8, 9 |
1110 |
|
GenBank mRNA |
AB033284.1 |
- |
11, 12 |
1631 |
|
AF284422.1 |
- |
11, 12 |
1671 |
|
AK024420.1 |
- |
6, 7 |
2757 |
|
AK024421.1 |
- |
7, 8 |
2521 |
|
AK024466.1 |
- |
1, 2 |
2857 |
|
AK024494.1 |
- |
6, 7 |
2829 |
|
AK090458.1 |
- |
6, 7 |
2735 |
|
AK301411.1 |
- |
11, 12 |
1704 |
|
BC000154.2 |
- |
9, 10 |
1423 |
2900382 |
CA488831.1 |
- |
|
582 |
|
View Full Product Details