Gene Transcripts

Gene Symbol
Atad5
Entrez Gene ID
237877
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
C130052G03Rik, FRAG1, Gm17
Chromosome Location
Chr. 11: 80089379 - 80135797 on Build GRCm38
UniGene ID
Mm.89551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029856.2 NP_001025027.1 11 3512
GenBank mRNA AY557610.1 - 11 3503
BC046463.1 - 7 809
BC141180.1 - 10 3008 9055792

View Full Product Details