Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
18636
Gene Name
complement factor properdin
Gene Aliases
BCFG, Pfc
Chromosome Location
Chr. X: 20925535 - 20931525 on Build GRCm38
UniGene ID
Mm.3064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008823.3 NP_032849.2 9 1420
GenBank mRNA AK150212.1 - 9 1381
AK152782.1 - 9 1371
AK154306.1 - 9 1405
AK159291.1 - 9 1421
AK159609.1 - 9 1370
AK159853.1 - 9 1370
AK171374.1 - 9 1404
BC132369.1 - 9 1404 40130646
BC138306.1 - 9 1404 8861326
X12905.1 - 8 1269

View Full Product Details