Gene Transcripts
- Gene Symbol
- Cd27
- Entrez Gene ID
- 21940
- Gene Name
- CD27 antigen
- Gene Aliases
- S152, Tnfrsf7, Tp55
- Chromosome Location
- Chr. 6: 125232622 - 125237080 on Build GRCm38
- UniGene ID
- Mm.367714
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033126.2 |
NP_001028298.1
|
5, 6 |
764 |
|
NM_001042564.1 |
NP_001036029.1
|
3, 4 |
452 |
|
NM_001286753.1 |
NP_001273682.1
|
4, 5 |
584 |
|
GenBank mRNA |
AI326735.1 |
- |
3, 4 |
452 |
|
AK153895.1 |
- |
5, 6 |
703 |
|
AK154005.1 |
- |
5, 6 |
748 |
|
BC139125.1 |
- |
5, 6 |
729 |
8862147 |
BC139126.1 |
- |
5, 6 |
729 |
8862148 |
BC171931.1 |
- |
3, 4 |
417 |
9054107 |
BE631582.1 |
- |
4, 5 |
403 |
|
DT924276.1 |
- |
4, 5 |
510 |
|
L24495.1 |
- |
5, 6 |
806 |
|
View Full Product Details