Gene Transcripts

Gene Symbol
Rdh8
Entrez Gene ID
235033
Gene Name
retinol dehydrogenase 8
Gene Aliases
Gm182, prRDH
Chromosome Location
Chr. 9: 20818504 - 20826163 on Build GRCm38
UniGene ID
Mm.297661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030290.1 NP_001025461.1 6 743

View Full Product Details