Gene Transcripts
- Gene Symbol
- Ott
- Entrez Gene ID
- 18422
- Gene Name
- ovary testis transcribed
- Gene Aliases
- AV272316
- Chromosome Location
- Chr. X: 149097952 - 149138152 on Build GRCm38
- UniGene ID
- Mm.420007
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X96604.1 |
- |
|
501 |
|
- Gene Symbol
- Gm10439
- Entrez Gene ID
- 382243
- Gene Name
- predicted gene 10439
- Gene Aliases
- ENSMUSG00000072923
- Chromosome Location
- Chr. X: 149594601 - 149636621 on Build GRCm38
- UniGene ID
- Mm.304331
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC109132.1 |
- |
4 |
776 |
|
View Full Product Details