Gene Transcripts

Gene Symbol
PSG5
Entrez Gene ID
5673
Gene Name
pregnancy specific beta-1-glycoprotein 5
Gene Aliases
FL-NCA-3, PSG
Chromosome Location
Chr. 19: 43167743 - 43186536 on Build GRCh38
UniGene ID
Hs.654415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130014.1 NP_001123486.1 2 226
NM_002781.3 NP_002772.3 2 226
XM_011527132.2 XP_011525434.1 2 224
XM_017027003.1 XP_016882492.1 2 224
GenBank mRNA BC012607.1 - 2 193
BG622527.1 - 2 194
DA835167.1 - 2 226
M25384.1 - 2 194
M73713.1 - 2 180

View Full Product Details