Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017928.3 | NP_001017928.1 | 3, 4 | 330 | |
NM_001308326.1 | NP_001295255.1 | 3, 4 | 356 | ||
XM_011512410.2 | XP_011510712.1 | 3, 4 | 308 | ||
GenBank mRNA | BC054051.1 | - | 3, 4 | 321 | |
BC062725.1 | - | 3, 4 | 330 | 6495960 | |
BC109378.1 | - | 3, 4 | 314 | ||
CD683826.1 | - | 3, 4 | 352 | ||
DA025111.1 | - | 3, 4 | 352 |