Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206646.1 | NP_001193575.1 | 2 | 152 | |
NM_001206647.1 | NP_001193576.1 | 2 | 152 | ||
NM_001206648.1 | NP_001193577.1 | 2 | 152 | ||
NM_020642.3 | NP_065693.2 | 2 | 152 | ||
NR_045417.1 | 1 | 334 | |||
NR_045418.1 | 1 | 334 | |||
XM_017018011.1 | XP_016873500.1 | 2 | 172 | ||
XM_017018012.1 | XP_016873501.1 | 2 | 180 | ||
GenBank mRNA | AF493783.1 | - | 2 | 152 | |
AF493784.1 | - | 2 | 152 | ||
AF493785.1 | - | 2 | 152 | ||
AK075308.1 | - | 271 | |||
AK294549.1 | - | 1 | 313 | ||
BC030996.2 | - | 1 | 82 | 4609949 | |
BP355293.1 | - | 1 | 334 |