Gene Transcripts

Gene Symbol
ABO
Entrez Gene ID
28
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Gene Aliases
A3GALNT, A3GALT1, GTB, NAGAT
Chromosome Location
Chr. 9: 133255176 - 133275214 on Build GRCh38
UniGene ID
Hs.654423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020469.2 NP_065202.2 7 807
GenBank mRNA AF134412.1 - 7 782
AF134413.1 - 7 782
AF134414.1 - 7 782
AF134415.1 - 7 781
AF134416.1 - 7 781
AK302203.1 - 6 655
AY579471.1 - 1 408
AY727862.1 - 2 543
BC069595.1 - 7 782 7262430
BC069814.1 - 7 782
BC111575.1 - 7 782 40069575
DQ139865.1 - 7 782
FR691751.1 - 7 782
FR691752.1 - 7 782
J05175.1 - 7 779
JN791441.1 - 7 782
JX398330.1 - 7 782
JX398331.1 - 7 782
JX398332.1 - 7 782
JX398333.1 - 7 782
JX519569.1 - 7 782
JX519570.1 - 7 782
KT282967.1 - 7 782

View Full Product Details