Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020655.3 | NP_065706.2 | 2 | 1273 | |
NR_073379.2 | 2 | 727 | |||
GenBank mRNA | AB042636.1 | - | 2 | 1182 | |
AK055486.1 | - | 1 | 84 | ||
AK295518.1 | - | 2 | 727 | ||
BC036533.2 | - | 2 | 1255 | 5264853 |