Gene Transcripts

Gene Symbol
NUMA1
Entrez Gene ID
4926
Gene Name
nuclear mitotic apparatus protein 1
Gene Aliases
NMP-22, NUMA
Chromosome Location
Chr. 11: 72002864 - 72080693 on Build GRCh38
UniGene ID
Hs.325978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286561.1 NP_001273490.1 6 588
NM_006185.3 NP_006176.2 5 490
NR_104476.1 5 409
XM_006718564.1 XP_006718627.1 4 305
XM_011545054.1 XP_011543356.1 4 305
XM_011545055.1 XP_011543357.1 5 483
XM_011545056.2 XP_011543358.1 6 477
XM_011545057.2 XP_011543359.1 6 467
XM_011545058.1 XP_011543360.1 4 504
XM_011545059.2 XP_011543361.1 6 469
XM_011545060.2 XP_011543362.1 6 463
XM_011545061.2 XP_011543363.1 4 323
XM_011545062.2 XP_011543364.1 4 336
XM_011545063.2 XP_011543365.1 4 303
XM_011545064.1 XP_011543366.1 4 290
XM_011545065.1 XP_011543367.1 4 305
XM_011545066.1 XP_011543368.1 4 305
XM_017017830.1 XP_016873319.1 5 345
XM_017017831.1 XP_016873320.1 4 341
GenBank mRNA AB210007.1 - 5 291
AB621824.1 - 6 472
AK290509.1 - 4 305
AK307577.1 - 5 308
AK309041.1 - 4 312
BC004165.2 - 5 324
BC027493.1 - 5 291
BC043499.1 - 5 291
BC103765.1 - 5 409
Z11583.1 - 5 308
Z11584.1 - 6 410

View Full Product Details