Gene Transcripts

Gene Symbol
Kcnk12
Entrez Gene ID
64119
Gene Name
potassium channel, two pore domain subfamily K, member 12
Gene Aliases
THIK-2
Chromosome Location
Chr. 6: 11373917 - 11374942 on Build Rnor_6.0
UniGene ID
Rn.64577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022292.1 NP_071628.1 1336
GenBank mRNA AF287300.1 - 1336

View Full Product Details