Gene Transcripts

Gene Symbol
Irx1
Entrez Gene ID
16371
Gene Name
Iroquois related homeobox 1 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 13: 71958232 - 71963723 on Build GRCm38
UniGene ID
Mm.316056
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010573.2 NP_034703.2 2 915
GenBank mRNA AF165984.1 - 2 680
AK133937.1 - 2 915

View Full Product Details