Gene Transcripts

Gene Symbol
Klk1b27
Entrez Gene ID
16619
Gene Name
kallikrein 1-related peptidase b27
Gene Aliases
Gk27, Klk21l, Klk27, mGK-27
Chromosome Location
Chr. 7: 44052290 - 44056712 on Build GRCm38
UniGene ID
Mm.445911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020268.3 NP_064664.1 3, 4 549
GenBank mRNA AB039275.1 - 3, 4 551
BC132090.1 - 3, 4 526 40130367
BC132665.1 - 3, 4 526 40130942

View Full Product Details