Gene Transcripts

Gene Symbol
Nipal3
Entrez Gene ID
74552
Gene Name
NIPA-like domain containing 3
Gene Aliases
9130020G22Rik, AI480660, Npal3
Chromosome Location
Chr. 4: 135447136 - 135494806 on Build GRCm38
UniGene ID
Mm.26548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028995.3 NP_083271.1 12 2399
GenBank mRNA AK049696.1 - 12 2433
AK138843.1 - 12 2400
AK148206.1 - 12 2422

View Full Product Details