Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178908.3 | NP_849239.2 | 2 | 1397 | |
GenBank mRNA | AK052861.1 | - | 1365 | ||
AK078850.1 | - | 2 | 1366 | ||
AK085925.1 | - | 2 | 1399 | ||
AK161070.1 | - | 2 | 1329 | ||
BC025893.1 | - | 2 | 1372 | 5031155 |