Gene Transcripts
- Gene Symbol
- Sly
- Entrez Gene ID
- 382301
- Gene Name
- Sycp3 like Y-linked
- Gene Aliases
- Chromosome Location
- Chr. Y: 57187206 - 57213358 on Build GRCm38
- UniGene ID
- Mm.304354
- Species
- Mus musculus
- Gene Symbol
- Gm20858
- Entrez Gene ID
- 100041033
- Gene Name
- Sycp3 like Y-linked pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Y: 25727935 - 25753940 on Build GRCm38
- UniGene ID
- Mm.359525
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AV269226.1 |
- |
|
13 |
|
View Full Product Details