Gene Transcripts

Gene Symbol
ZNF14
Entrez Gene ID
7561
Gene Name
zinc finger protein 14
Gene Aliases
GIOT-4, KOX6
Chromosome Location
Chr. 19: 19710472 - 19733112 on Build GRCh38
UniGene ID
Hs.659932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021030.2 NP_066358.2 2 264
GenBank mRNA AB021644.1 - 2 214
AK074832.1 - 2 249
BC136323.1 - 2 151 9020310
BM836276.1 - 2 264

View Full Product Details