Gene Transcripts
- Gene Symbol
- CYP2A7
- Entrez Gene ID
- 1549
- Gene Name
- cytochrome P450 family 2 subfamily A member 7
- Gene Aliases
- CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4
- Chromosome Location
- Chr. 19: 40875439 - 40885952 on Build GRCh38
- UniGene ID
- Hs.719890
- Species
- Homo sapiens
- Gene Symbol
- CYP2A7P1
- Entrez Gene ID
- 1550
- Gene Name
- cytochrome P450 family 2 subfamily A member 7 pseudogene 1
- Gene Aliases
- CYP2A18PC, CYP2A18PN, CYP2A7P2, CYP2A7PT
- Chromosome Location
- Chr. 19: 41024251 - 41028263 on Build GRCh38
- UniGene ID
- Hs.733030
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U22030.1 |
- |
1 |
527 |
|
U22044.1 |
- |
1 |
527 |
|
View Full Product Details