Gene Transcripts

Gene Symbol
CXCL6
Entrez Gene ID
6372
Gene Name
C-X-C motif chemokine ligand 6
Gene Aliases
CKA-3, GCP-2, GCP2, SCYB6
Chromosome Location
Chr. 4: 73836556 - 73838760 on Build GRCh38
UniGene ID
Hs.164021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002993.3 NP_002984.1 4 960
GenBank mRNA BC013744.1 - 4 795
U81234.1 - 4 828

View Full Product Details