Gene Transcripts

Gene Symbol
ZNF14
Entrez Gene ID
7561
Gene Name
zinc finger protein 14
Gene Aliases
GIOT-4, KOX6
Chromosome Location
Chr. 19: 19710472 - 19733112 on Build GRCh38
UniGene ID
Hs.659932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021030.2 NP_066358.2 3 312
GenBank mRNA AB021644.1 - 3 262
AK074832.1 - 3 297
BC136323.1 - 3 199 9020310
BM836276.1 - 3 312

View Full Product Details