Gene Transcripts

Gene Symbol
PCDHB8
Entrez Gene ID
56128
Gene Name
protocadherin beta 8
Gene Aliases
PCDH-BETA8, PCDH3I
Chromosome Location
Chr. 5: 141177790 - 141180500 on Build GRCh38
UniGene ID
Hs.283803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019120.4 NP_061993.3 1 1106
GenBank mRNA AF152501.2 - 1 861
BC136801.1 - 1 1106 9020791

View Full Product Details