Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191013.1 | NP_001177942.1 | 4 | 794 | |
NM_001191014.1 | NP_001177943.1 | 2 | 229 | ||
NM_001191015.1 | NP_001177944.1 | 2 | 229 | ||
NM_183239.1 | NP_899062.1 | 4 | 794 | ||
XM_006717624.3 | XP_006717687.1 | 4 | 483 | ||
XM_011539270.2 | XP_011537572.1 | 4 | 1857 | ||
XM_011539271.1 | XP_011537573.1 | 4 | 483 | ||
XM_011539272.2 | XP_011537574.1 | 4 | 483 | ||
XM_011539273.2 | XP_011537575.1 | 4 | 483 | ||
XM_017015670.1 | XP_016871159.1 | 4 | 483 | ||
XM_017015671.1 | XP_016871160.1 | 4 | 483 | ||
GenBank mRNA | AK291886.1 | - | 4 | 482 | |
AK296266.1 | - | 3 | 181 | ||
AK297522.1 | - | 2 | 192 | ||
AK300508.1 | - | 3 | 191 | ||
AY191318.1 | - | 3 | 162 | ||
AY209189.1 | - | 4 | 486 | ||
AY350731.1 | - | 4 | 486 | ||
BC023583.2 | - | 2 | 195 | ||
BC056918.1 | - | 4 | 794 | 5165084 | |
BM708208.1 | - | 2 | 133 | ||
DC341647.1 | - | 3 | 181 |