Gene Transcripts

Gene Symbol
DNAJC12
Entrez Gene ID
56521
Gene Name
DnaJ heat shock protein family (Hsp40) member C12
Gene Aliases
JDP1
Chromosome Location
Chr. 10: 67796665 - 67838179 on Build GRCh38
UniGene ID
Hs.260720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021800.2 NP_068572.1 1 199
NM_201262.1 NP_957714.1 1 199
XM_017016432.1 XP_016871921.1 1 21
GenBank mRNA AF176012.1 - 1 184
AF176013.1 - 1 116
AK289537.1 - 1 149
BC017018.1 - 1 135 3884093
CB121724.1 - 1 201
CR457184.1 - 1 31

View Full Product Details