Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290185.1 | NP_001277114.1 | 2 | 491 | |
NM_021729.5 | NP_068375.3 | 2 | 276 | ||
XM_011542899.2 | XP_011541201.1 | 2 | 234 | ||
GenBank mRNA | AB027508.2 | - | 2 | 246 | |
AF308800.1 | - | 2 | 210 | ||
AK023279.1 | - | 2 | 248 | ||
AK056701.1 | - | 2 | 239 | ||
AK223158.1 | - | 2 | 236 | ||
AK302973.1 | - | 2 | 423 | ||
AL834218.1 | - | 2 | 225 | ||
BC065563.1 | - | 2 | 210 | 6144541 | |
BI561271.1 | - | 2 | 281 |