Gene Transcripts

Gene Symbol
ADAM30
Entrez Gene ID
11085
Gene Name
ADAM metallopeptidase domain 30
Gene Aliases
svph4
Chromosome Location
Chr. 1: 119893533 - 119896524 on Build GRCh38
UniGene ID
Hs.283011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021794.3 NP_068566.2 1 499
GenBank mRNA AF171932.1 - 1 311
AF171933.1 - 1 311
AK292483.1 - 1 490
AY358734.1 - 1 473
BC028372.2 - 1 470 4831258

View Full Product Details