Gene Transcripts

Gene Symbol
UBE2O
Entrez Gene ID
63893
Gene Name
ubiquitin conjugating enzyme E2 O
Gene Aliases
E2-230K
Chromosome Location
Chr. 17: 76389532 - 76453206 on Build GRCh38
UniGene ID
Hs.16130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022066.3 NP_071349.3 14 2218
XM_005257567.1 XP_005257624.1 14 2205
XM_017024934.1 XP_016880423.1 14 2130
XM_017024935.1 XP_016880424.1 14 2127
GenBank mRNA AB051521.1 - 14 2218
AK025999.1 - 6 697
AK225412.1 - 6 697
AL832432.1 - 7 1143
BC036820.1 - 9 1354
BC051868.2 - 6 561 6162673

View Full Product Details