Gene Transcripts
- Gene Symbol
- PBLD
- Entrez Gene ID
- 64081
- Gene Name
- phenazine biosynthesis like protein domain containing
- Gene Aliases
- HEL-S-306, MAWBP, MAWDBP
- Chromosome Location
- Chr. 10: 68282660 - 68332958 on Build GRCh38
- UniGene ID
- Hs.198158
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033083.1 |
NP_001028255.1
|
6, 7 |
622 |
|
NM_022129.3 |
NP_071412.2
|
6, 7 |
622 |
|
XM_005270028.3 |
XP_005270085.1
|
6, 7 |
656 |
|
XM_011540060.2 |
XP_011538362.1
|
6, 7 |
656 |
|
XM_017016513.1 |
XP_016872002.1
|
6, 7 |
615 |
|
GenBank mRNA |
AB049758.1 |
- |
4, 5 |
484 |
|
AI114748.1 |
- |
3, 4 |
157 |
|
AK027673.1 |
- |
6, 7 |
590 |
|
AK092826.1 |
- |
4, 5 |
355 |
|
AK312275.1 |
- |
5, 6 |
413 |
|
BC009738.1 |
- |
6, 7 |
604 |
3879159 |
GQ472233.1 |
- |
6, 7 |
604 |
|
View Full Product Details