Gene Transcripts
- Gene Symbol
- CHST8
- Entrez Gene ID
- 64377
- Gene Name
- carbohydrate sulfotransferase 8
- Gene Aliases
- GALNAC4ST1, GalNAc4ST, PSS3
- Chromosome Location
- Chr. 19: 33621955 - 33773509 on Build GRCh38
- UniGene ID
- Hs.165724
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127895.1 |
NP_001121367.1
|
3, 4 |
636 |
|
NM_001127896.1 |
NP_001121368.1
|
2, 3 |
547 |
|
NM_022467.3 |
NP_071912.2
|
2, 3 |
889 |
|
XM_011527222.1 |
XP_011525524.1
|
2, 3 |
870 |
|
XM_011527224.1 |
XP_011525526.1
|
3, 4 |
1004 |
|
XM_017027143.1 |
XP_016882632.1
|
2, 3 |
540 |
|
GenBank mRNA |
AB047801.1 |
- |
2, 3 |
403 |
|
AF300612.1 |
- |
2, 3 |
350 |
|
AF305781.1 |
- |
2, 3 |
446 |
|
BC011380.1 |
- |
2, 3 |
499 |
3874578 |
BC014250.2 |
- |
2, 3 |
543 |
4635405 |
BC018723.1 |
- |
2, 3 |
889 |
4137487 |
BI917117.1 |
- |
2, 3 |
547 |
|
BX354555.2 |
- |
3, 4 |
635 |
|
View Full Product Details