Gene Transcripts
- Gene Symbol
- TM6SF1
- Entrez Gene ID
- 53346
- Gene Name
- transmembrane 6 superfamily member 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 83107572 - 83145403 on Build GRCh38
- UniGene ID
- Hs.513094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001144903.1 |
NP_001138375.1
|
6, 7 |
720 |
|
NM_023003.3 |
NP_075379.2
|
6, 7 |
720 |
|
XM_011521679.2 |
XP_011519981.1
|
6, 7 |
645 |
|
XM_011521680.2 |
XP_011519982.1
|
6, 7 |
645 |
|
XM_017022320.1 |
XP_016877809.1
|
6, 7 |
645 |
|
XM_017022321.1 |
XP_016877810.1
|
6, 7 |
645 |
|
XM_017022322.1 |
XP_016877811.1
|
6, 7 |
645 |
|
XM_017022323.1 |
XP_016877812.1
|
6, 7 |
772 |
|
XM_017022324.1 |
XP_016877813.1
|
6, 7 |
749 |
|
XM_017022325.1 |
XP_016877814.1
|
6, 7 |
634 |
|
XM_017022326.1 |
XP_016877815.1
|
6, 7 |
634 |
|
XM_017022327.1 |
XP_016877816.1
|
6, 7 |
826 |
|
GenBank mRNA |
AF255922.1 |
- |
6, 7 |
668 |
|
AK223246.1 |
- |
6, 7 |
665 |
|
AK292100.1 |
- |
6, 7 |
645 |
|
AK295568.1 |
- |
6, 7 |
609 |
|
View Full Product Details