Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
Chromosome Location
Chr. 17: 81977550 - 82018929 on Build GRCh38
UniGene ID
Hs.298351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251888.1 NP_001238817.1 4 420
NM_024083.3 NP_076988.1 4 420
NR_045351.1 3 364
XM_011523601.2 XP_011521903.1 4 420
XM_011523602.2 XP_011521904.1 4 420
XM_017025036.1 XP_016880525.1 4 420
XM_017025037.1 XP_016880526.1 4 420
XM_017025038.1 XP_016880527.1 5 431
XM_017025039.1 XP_016880528.1 5 348
XM_017025040.1 XP_016880529.1 4 285
XM_017025041.1 XP_016880530.1 3 288
XM_017025042.1 XP_016880531.1 4 420
XM_017025043.1 XP_016880532.1 3 288
XM_017025044.1 XP_016880533.1 5 431
GenBank mRNA AF324219.2 - 4 345
AK057403.1 - 4 420
AK057851.1 - 3 272
AK226086.1 - 4 293
AK290624.1 - 4 336
BC006152.1 - 4 375 3357489
BC018722.1 - 4 325 4640076

View Full Product Details