Gene Transcripts
- Gene Symbol
- ASPSCR1
- Entrez Gene ID
- 79058
- Gene Name
- ASPSCR1, UBX domain containing tether for SLC2A4
- Gene Aliases
- ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
- Chromosome Location
- Chr. 17: 81977550 - 82018929 on Build GRCh38
- UniGene ID
- Hs.298351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001251888.1 |
NP_001238817.1
|
6, 7 |
611 |
|
NM_024083.3 |
NP_076988.1
|
6, 7 |
611 |
|
NR_045351.1 |
|
5, 6 |
555 |
|
XM_011523601.2 |
XP_011521903.1
|
6, 7 |
611 |
|
XM_011523602.2 |
XP_011521904.1
|
6, 7 |
611 |
|
XM_017025036.1 |
XP_016880525.1
|
6, 7 |
611 |
|
XM_017025037.1 |
XP_016880526.1
|
6, 7 |
611 |
|
XM_017025038.1 |
XP_016880527.1
|
7, 8 |
622 |
|
XM_017025039.1 |
XP_016880528.1
|
7, 8 |
539 |
|
XM_017025040.1 |
XP_016880529.1
|
6, 7 |
476 |
|
XM_017025041.1 |
XP_016880530.1
|
5, 6 |
479 |
|
XM_017025042.1 |
XP_016880531.1
|
6, 7 |
611 |
|
XM_017025043.1 |
XP_016880532.1
|
5, 6 |
479 |
|
XM_017025044.1 |
XP_016880533.1
|
7, 8 |
622 |
|
GenBank mRNA |
AF324219.2 |
- |
6, 7 |
536 |
|
AK057403.1 |
- |
6, 7 |
611 |
|
AK057851.1 |
- |
5, 6 |
463 |
|
AK226086.1 |
- |
6, 7 |
484 |
|
AK290624.1 |
- |
6, 7 |
527 |
|
BC006152.1 |
- |
6, 7 |
566 |
3357489 |
BC018722.1 |
- |
6, 7 |
516 |
4640076 |
View Full Product Details