Gene Transcripts
- Gene Symbol
- SCRN3
- Entrez Gene ID
- 79634
- Gene Name
- secernin 3
- Gene Aliases
- SES3
- Chromosome Location
- Chr. 2: 174395729 - 174430071 on Build GRCh38
- UniGene ID
- Hs.470679
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193528.1 |
NP_001180457.1
|
7, 8 |
1107 |
|
NM_024583.4 |
NP_078859.2
|
7, 8 |
1198 |
|
XM_005246853.2 |
XP_005246910.1
|
7, 8 |
1603 |
|
XM_005246854.4 |
XP_005246911.1
|
7, 8 |
2763 |
|
XM_005246855.2 |
XP_005246912.1
|
7, 8 |
1651 |
|
XM_005246856.2 |
XP_005246913.1
|
7, 8 |
1607 |
|
XM_005246857.2 |
XP_005246914.1
|
7, 8 |
1307 |
|
XM_011511839.2 |
XP_011510141.1
|
6, 7 |
1007 |
|
XM_017004909.1 |
XP_016860398.1
|
7, 8 |
2195 |
|
XM_017004910.1 |
XP_016860399.1
|
7, 8 |
1673 |
|
GenBank mRNA |
AF279776.1 |
- |
3, 4 |
535 |
|
AK026795.1 |
- |
3, 4 |
545 |
|
AK057912.1 |
- |
7, 8 |
1176 |
|
AK295357.1 |
- |
7, 8 |
1091 |
|
AK296558.1 |
- |
6, 7 |
997 |
|
BC031821.1 |
- |
4, 5 |
659 |
|
BC119684.1 |
- |
7, 8 |
1149 |
40116568 |
BC119685.1 |
- |
7, 8 |
1149 |
40116570 |
View Full Product Details